Learn More
BERTECH KELEX Industrial Grade Finger Cots, Ultra Tough, 14 Mil Thick, Medium (300 Pcs./Bag)
Industrial Grade Finger Cots are made of heavy 14 mil natural rubber latex material. These finger cots are tough, durable, and re-usable. They are primarily used where sharp objects are involved. These heavy duty finger cots are unrolled, lightly powdered, and textured for a better grip. They are packaged as 300 pieces per bag.Applications: Automotive, Banking, Electronic Assembly, Industrial and retail facilities.