Learn More
TARGETMOL CHEMICALS INC ORDOPIDINE 5MG
Supplier:  TARGETMOL CHEMICALS INC T33816
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Ordopidine (ACR325) is a dopaminergic stabilizer that suppresses psychostimulant-induced hyperactivity disorder and stimulates behavior during inactivity. Ordopidine acts as a dopamine D2 receptor antagonist in vitro and despite its low affinity its specific state-dependent behavioral effect characteristics are not generally shared by D2 receptor antagonists. purity: 99%
Catalog No. 50-258-0291
missing translation for 'QuantityLeft'