Learn More
Eurofins Discoverx Orexin A, 1 mg. (92-1123)
Supplier:  Eurofins Discoverx 921123
Encompass
Orexin A is a neuropeptide hormone released by the hypothalamus that excites neurons and affects hunger and wakefulness by modulating the dopamine, norepinephrine, histamine, and acetylcholine systems. Lack of orexin A tends to result in narcolepsy. Potent orexin receptor agonist (Ki=20 and 38 nM for OX1 and OX2, respectively). Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9656
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.