Learn More
Perkin Elmer US LLC Organics Sample Introduction Torch Module for Avio 500
Supplier:  Perkin Elmer US LLC N0810609
Encompass_Preferred
This kit includes: GemCone Low-Flow Nebulizer (N0770358) Polyethylene Tubing for Nebulizer, 10 ft (09908265) Cyclonic Injector Adaptor Kit (N0790141) 3-Slot HybridXLT Organics Quartz Torch (N0790249) 1.2 mm I.D. Alumina Injector (N0791182) Glass Baffled Cyclonic Spray Chamber (N0791352) Adjustable Torch Body Assembly (N0810128)
Catalog No. 50-211-3685
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.