Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organometallic Oil Standard, 21-Element For Wear Metal Analysis, SPEX CertiPrep™
SDP

Catalog No. S21900Z Shop All Spex CertiPrep Products
Click to view available options
Quantity:
Packaging:

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag900μg/g
Al900μg/g
B900μg/g
Ba900μg/g
Ca900μg/g
Cd900μg/g
Cr900μg/g
Cu900μg/g
Fe900μg/g
Mg900μg/g
Mn900μg/g
Mo900μg/g
Na900μg/g
Ni900μg/g
P900μg/g
Pb900μg/g
Si900μg/g
Sn900μg/g
Ti900μg/g
V900μg/g
Zn900μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material

21 Element Oil Standard

Packaging

Triple-leached LDPE bottle

Quantity

50 g

Certificate of Analysis

With Certificate of Analysis.

Concentration or Composition Notes

Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.

For Use With (Equipment)

AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation

Grade

Oil

Solvent or Matrix

Base Oil 75

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.