Learn More
Electron Microscopy Sciences Oriented Single Crystal Gold
Supplier: Electron Microscopy Sciences 80038
Since crystal lattice plane spacings are accurately known from x-ray measurements, they provide a good test of microscope stability and a calibration of magnification in the upper range of magnification of the instrument. Plane Spacing 0.204 nm, 0.143 nm and 0.102 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.