Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ All-in-One pH Buffer Kit PROMO

Catalog No. 0935772PM Shop All Thermo Scientific Products
Click to view available options
:
Quantity: 5 x 60 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Perform accurate, reproducible pH calibrations with color-coded Thermo Scientific™ Orion™ pH Buffer Bottles. pH 4.01, 7.00 and 10.01 buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.

All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability, portable and field analysis. Manufactured under ISO 9000 quality standards.

Buffer Color Codes:

  • pH 4.01 buffer is pink
  • pH 7.00 buffer is yellow
  • pH 10.01 buffer is blue

Buffers are available in pint (475 mL) bottles, portable five packs of 2 oz. (5 x 60 mL) bottles, convenient All-in-One buffer kits, 5 gallon (19 L) Cubitainers™ and single-use pouches.

Note: Image is representative only. See specifications for which buffers and solutions are included in this promo package.

Specifications

Description Small Bottle All-in-One pH Buffer Kit
For Use With (Application) Calibration
pH 4.01, 7.00, 10.01
Quantity 5 x 60 mL
Packaging Kit
Traceability to NIST Yes
Includes 60 mL bottle each of pH 4.01, 7.00 and 10.01 buffers, 60 mL bottle of pH electrode storage solution, and 60 mL bottle of pH electrode cleaning solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.