Learn More
ORP150/HSP12A Antibody (6G7-2H5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259346
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ORP150/HSP12A | |
Monoclonal | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.1% Sodium Azide | |
HYOU1 | |
Raised against a synthetic peptide of human GRP170 | |
100 μg | |
Hypoxia | |
10525 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6G7-2H5 | |
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects 170kDa. | |
Store at -20C. | |
IgG2a |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only