Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ B12/Folate Denaturing Reagent

Supplier: Ortho-Clinical Diagnostics™ 1142561
Specifications
Vitamin B12 | |
B12/Folate Denaturing Reagent | |
11.7mL Sample treatment (denaturation) reagent, 15.6mL sample treatment (neutralization) reagent, ancillary lot card |
EDTA Plasma, Heparin Plasma, Serum | |
Folate deficiency, in-vitro diagnostic use only | |
200 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction