Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Anti-Hepatitis C Virus
SDP
Supplier:  Ortho-Clinical Diagnostics™ 6801326
Catalog No. 23-057-293
Item Discontinued This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Explore all special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
2°C to 8°C
Anti-HCV Calibrator
Qualitative detection of Immunoglobulin G Antibody to Hepatitis C Virus (Anti-HCV)
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system
2mL Anti-HCV calibrator (Anti-HCV positive Human Plasma in Anti-HCV negative Human Plasma with antimicrobial agent), lot calibration card, protocol card, 8 calibrator bar code labels
1 Set/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.