Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Anti-Hepatitis C virus (A-HCV)

Supplier: Ortho-Clinical Diagnostics™ 6801325
This item is not returnable.
View return policy
Specifications
Hepatitis C Virus IgG | |
Vitros™ Anti-HCV Reagent | |
100 Pack |
Well | |
Coated wells 100, assay reagent 18.2mL, conjugate reagent 20.6mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction