Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Rubella IGG

Supplier: Ortho-Clinical Diagnostics™ 8536195
This item is not returnable.
View return policy
Specifications
Rubella IgG | |
Vitros™ Rubella IgG Reagent | |
100 Pack | |
35 min. |
Well | |
Coated wells 100, assay reagent 19.4mL, conjugate reagent 20.6mL | |
10 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction