Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Total B-HCG II

Supplier: Ortho-Clinical Diagnostics™ 6802220
This item is not returnable.
View return policy
Specifications
Human chorionic gonadotropin | |
Vitros™ Total Beta-hCG II Reagent | |
100 Pack | |
24 min. |
Well | |
Coated wells 100, Biotinylated Antibody reagent 14.4mL, conjugate reagent 19.2mL | |
40 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction