Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium 99.9%, Thermo Scientific Chemicals

Catalog No. p-3523531
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 7440-04-2
Molecular Formula Os
Molecular Weight (g/mol) 190.23
MDL Number MFCD00011147
InChI Key SYQBFIAQOQZEGI-UHFFFAOYSA-N
Synonym ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium
SMILES [Os]

Specifications

Color Black
Melting Point 3045.0°C
Boiling Point 5027.0°C
Assay Percent Range 99.90%
Linear Formula Os
Quantity 1 g
Merck Index 15, 6988
Solubility Information Solubility in water: insoluble.
Formula Weight 190.2
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Form Powder
Chemical Name or Material Osmium
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.