Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Ovation Electronic Multichannel (E12) Pipette
SDP

Catalog No. p-200025622 Shop All VistaLab Technologies Inc. Products
Click to view available options
Volume (Metric):
For Use With (Equipment):

Ovation™ pipettes have a unique and patented ergonomic design. They are the only pipettes that fully address the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting, while always providing superior accuracy and precision.

  • The angled shape allows users to pipette with low arm and elbow height, decreasing tension and associated pain in the neck, shoulder, back, and elbow. When used properly, the Ovation™ pipette will keep the user's hand and wrist in the neutral position recommended by ergonomic experts.
  • Contoured body shape and adjustable hook assures a comfortable, easy grip, and custom fit hand hold spring-loaded tip acquisition means a gentle push is all that is required to attach tips, and an audible -click- confirms secure attachment.
  • Tips -flick- off easily with a low force push of the eject button.
  • Special filters in the nozzle prevent liquid from being inadvertently aspirated into the pipettor, protecting it from contamination.
  • Ovation™ pipettes are the only pipettes that stand on their own, which means tips never touch the bench, even if the pipette is put down in the middle of a procedure.
  • Mechanical models feature two-stroke operation, available with -click stop- or -quick set digital- volume adjustment (0.2 μL to 1000 μL)
  • Electronic models feature custom volume presets, multi-dispensing, mixing, and serial diluting (0.5 μL to 1250 μL)

Specifications

Compatible Tips

LfTS 1000μL

Adjustable Tip Spacing

No

Autoclavable

Not Autoclavable

Color

Gray

Dispensing Speed

Variable

Disposable

Reusable

For Use With

Pipette Tips

Grip Type

Ergonomic

Material

PVDF

Number of Channels

12

Programmability

No

Sterility

Non-sterile

Type

Electronic Micropipette

Volume (Metric)

850 μL

For Use With (Equipment)

300 μL Pipette Tips

Color Code

Yes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.