Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid , For Silica, 5% (w/v), LabChem™

Catalog No. p-5388960
Click to view available options
Quantity:
500mL
1L

Chemical Identifiers

CAS 6153-56-6, 7732-18-5
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

CAS Max % 95, 5
Linear Formula HO2CCO2H
Formula Weight 90.04
Physical Form Liquid
Decomposition Information Carbon monoxide; Carbon dioxide; Formic acid
Percent Purity 5% (w/v)
Color Colorless
Quantity 500mL
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Chemical Name or Material Oxalic Acid, For Silica
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.