Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxfendazole 98.0+%, TCI America™
SDP

Catalog No. p-7134311 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15H13N3O3S
Molecular Weight (g/mol) 315.347
MDL Number MFCD00801063
InChI Key BEZZFPOZAYTVHN-UHFFFAOYSA-N
Synonym Methyl [5-(Phenylsulfinyl)benzimidazol-2-yl]carbamate, [5-(Phenylsulfinyl)benzimidazol-2-yl]carbamic Acid Methyl Ester
PubChem CID 40854
ChEBI CHEBI:35812
IUPAC Name methyl N-[6-(benzenesulfinyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)S(=O)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 315.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxfendazole
RTECSNumber FD0808000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.