Learn More
Cell Biolabs Inc OxiSelect™ Free Hydrogen Sulfide Gas Assay Kit, 100 assays
Supplier:  Cell Biolabs Inc XAN5084
Encompass_Preferred
Cell Biolabs' OxiSelect™ Free Hydrogen Sulfide Gas Assay Kit is a novel assay for the detection of hydrogen sulfide gas emission in a 96-well plate. As H2S gas is emitted from samples in the well of the plate, they are captured by a polymer matrix coated onto the plate lid. When the hydrogen sulfide gas enters the polymer matrix, a color is produced that can be quantified in a standard colorimetric microplate reader.
Catalog No. NC1652288
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.