Learn More
Uline OXYGEN ABSORBING PACKTS 2000/C
Supplier: Uline S23091
Oxygen-Absorbing Packets - 300 ccPreserve freshness and extend the shelf life of packaged foods.Prevents mold and bacteria growth.Removes over 99.9% of oxygen from sealed, airtight packages.PROTECTS 100 CU. IN.FDA compliant.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.