Learn More
Hach Company Oxygen Demand Quality Control Standard, pk/16 - 10 mL Voluette Ampule
Supplier:  Hach Company 2833510
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Ready to use - no dilutions required. Oxygen Demand Quality Control Standard. BOD - 396 mg/L. COD - 617 mg/L. TOC - 245 mg/L.
Catalog No. 50-199-6740
missing translation for 'QuantityLeft'