Learn More
Advanced Micro Instruments P-3 PERCENT, OXYGEN SENSOR
Supplier: Advanced Micro Instruments 4SEN04
AMI MODEL P-3 SENSORP-3 SENSOR0 TO 25 PERCENT O2 RANGEMINIMUM RANGE 0 TO 1000 PPM100 PERCENT CO2 RESISTANT
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.