Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dioxane, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391190 Shop All Avantor Products
Click to view available options
Quantity:
150 mL
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Formula Weight 88.11
Grade BAKER ANALYZED™ ACS Reagent
Quantity 150 mL
Flash Point 12.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.03kg
Chemical Name or Material p-Dioxane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.