Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals

Catalog No. p-201170
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207801-20-5
Molecular Formula C7H7NaO2S
Molecular Weight (g/mol) 178.18
MDL Number MFCD00149640
InChI Key KFZUDNZQQCWGKF-UHFFFAOYSA-M
Synonym sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water
PubChem CID 23682957
IUPAC Name sodium;4-methylbenzenesulfinate;hydrate
SMILES [Na+].CC1=CC=C(C=C1)S([O-])=O

Specifications

Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula CH3C6H4SO2Na·xH2O
Quantity 100 g
Beilstein 11, 9
Fieser 08,488
Merck Index 10, 9360
Solubility Information Solubility in water: soluble
Formula Weight 178.18
Percent Purity 98+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material p-Toluenesulfinic acid,sodium salt hydrate, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 212-538-5
RTECSNumber XT4725000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.