Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P16-INK4A Polyclonal Antibody, CoraLite555, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL55510883100UL

Description
P16-INK4A Polyclonal antibody specifically detects P16-INK4A in Human samples. It is validated for Flow CytometrySpecifications
P16-INK4A | |
Polyclonal | |
CoraLite 555 | |
P42771 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16 | |
Antigen affinity chromatography | |
RUO | |
1029 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction