Learn More
Enzo Life Sciences p300 (catalytic domain) (human), (recombinant) (100 µg)
Supplier: Enzo Life Sciences BMLSE4510100
Synonyms: E1A binding protein p300, Histone acetyltransferase p300. MW: 45.1 kDa. UniProt ID: Q09472. Source: Produced in E. coli. Catalytic domain (aa 1284-1673) of human p300. Purification: Partially purified by single-step affinity chromatography and gel filtration. Formulation: Liquid. In 50mM TRIS/HCl, pH 8.0, containing 0.1mM EDTA, 10% glycerol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.