Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P53 Recombinant Rabbit Monoclonal Antibody (2H13), CoraLite488, Proteintech

Rabbit Recombinant Monoclonal Antibody
Supplier: Proteintech Group Inc CL48880077100UL

Description
P53 Recombinant Monoclonal antibody specifically detects P53 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
P53 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
bbl; BCC7; bfy; bhy; LFS1; p44; P53; TP53; TRP53 | |
Protein A | |
RUO | |
7157 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Immunocytochemistry | |
2H13 | |
CoraLite 488 | |
P04637 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction