Learn More
Wilmad Labglass PACKING SOLID GLASS BEADS 5MM
Supplier: Wilmad Labglass LG6750104
Distillation Column Packing, Solid Glass Beads. Approximate Diameter: 5mm; Approximate Number Per Pound: 2,500. Manufactured from borosilicate glass. Sold in one pound packages.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.