Learn More
Medchemexpress LLC HY-B0015 500mg , Paclitaxel CAS:33069-62-4 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYB0015500MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-B0015 500mg Paclitaxel CAS:33069-62-4 Formula:C47H51NO14 IC50: 4 nM (MT) Purity:98% Paclitaxel is a potent anticancer agent known to promote microtubule (MT)  assembly, inhibit MT depolymerization, and change MT dynamics required for mitosis and cell proliferation. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2330