Learn More
Cell Biolabs Inc PAK1 PBD Agarose Beads, 400 µg
Supplier:  Cell Biolabs Inc STA411
Encompass_Preferred
Rac, a 21 kDa protein, belongs to the family of Rho GTPases regulating a variety of biological response pathways that include cell motility, cell division, gene transcription, and cell transformation. Like other small GTPases, Rac regulates molecular events by cycling between an inactive GDP-bound form and an active GTP-bound form. In its active (GTP-bound) state, Rac binds specifically to the p21-binding domain (PBD) of p21-activated protein kinase (PAK) to control downstream signaling cascades. PAK PBD Agarose beads (pink in color) are easy to visualize, minimizing potential loss during washes and aspirations of Rac-GTP pulldown
Catalog No. 50-673-615
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.