Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paliperidone, Tocris Bioscience™

Catalog No. p-4817263 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Atypical antipsychotic; 5-HT2A and D2 antagonist

Atypical antipsychotic; metabolite of risperidone (Cat. No. 2865). Exhibits high affinity for 5-HT2A receptors (Ki = 0.4 nM for cloned human 5-HT2A receptors). Also displays nanomolar affinity for human D2 receptors.

Chemical Identifiers

CAS 144598-75-4
Molecular Formula C23H27FN4O3
Molecular Weight (g/mol) 426.492
InChI Key PMXMIIMHBWHSKN-UHFFFAOYSA-N
Synonym paliperidone, 9-hydroxyrisperidone, invega, 9-oh-risperidone, invega sustenna, xeplion, palliperidone, 3-2-4-6-fluoro-1,2-benzisoxazol-3-yl-1-piperidinyl ethyl-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4h-pyrido 1,2-a pyrimidin-4-one, chembl1621, 3-2-4-6-fluorobenzo d isoxazol-3-yl piperidin-1-yl ethyl-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4h-pyrido 1,2-a pyrimidin-4-one
PubChem CID 115237
ChEBI CHEBI:83804
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCC(C2=N1)O)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Quantity 10 mg
Formula Weight 426.48
Percent Purity >98%
Chemical Name or Material Paliperidone
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.