Learn More
Electron Microscopy Sciences Palladium/Gold 40, 0.20 mm, Purity 99.90% 10 FT
Supplier:  Electron Microscopy Sciences 73400
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.90%
Catalog No. 50-280-55
missing translation for 'QuantityLeft'