Learn More
AdipoGen Palmarumycin C3
SDP
Supplier:  AdipoGen BVT0078M001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 159934-11-9. Formula C20H12O6. MW 348.3. Isolated from Sphaeropsidales sp. Rasfarnesyltransferase inhibitor. Antifungal, antibacterial and herbicidal. Structurally unique natural products. The basic structure of palmarumycin C1 can be modified by a number of hydroxylation, oxygenation, dehydrogenation and chlorination steps. Antioxidant
Catalog No. 50-256-2117
missing translation for 'QuantityLeft'