Learn More
Bioassay Systems Parallel Artificial Membrane Permeability Assay-BBB Kit. Assess blood brain barrier (BBB) membrane permeability of test compounds.
SDP
Supplier:  Bioassay Systems PMBBB096
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitative determination of blood brain barrier (BBB) membrane permeability of test compounds. Key Features: Convenient. Includes all necessary equipment to run a PAMPA plate. Simple and low-cost. Procedure is easy to follow and more affordable than cell-based permeability assays. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: PAMPA. Samples: Chemical compounds. Procedure: Assay takes 20 hr, hands-on time 1 hr. Kit size: 96 tests.
Catalog No. 50-225-5890