Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. p-7157928 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle

P1000, 85-85-8, C15H11N3O

Spectrum™ Chemical PAN, Reagent, ACS

Chemical Identifiers

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-SDXDJHTJSA-N
IUPAC Name (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.