Learn More
Enzo Life Sciences Paroxetine. HCl (100mg). CAS: 110429-35-1; 78246-49-8; 61869-08-7 (free base)
Supplier: Enzo Life Sciences BMLNS7100100
Paroxetine is a highly potent and selective serotonin (5-HT) uptake inhibitor. It acts by binding to the serotonin transporter (SERT) with high affinity (Ki = 05 nM). In vitro paroxetine competitively inhibits the uptake of 5-HT by rat hypothalamic and cortical synaptosomes with only very weak effects on noradrenaline and dopamine uptake. Paroxetine is in clinical use for the treatment of depression, anxiety and other disorders. Alternative name: Paxil. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.