Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Paroxetine maleate
Highly potent and selective 5-HT uptake inhibitor
$224.00 - $902.50
Specifications
Purity | >99% |
---|---|
Product Type | Paroxetine maleate |
Molecular Weight (g/mol) | 445.44 |
Inhibitors | Paroxetine maleate |
Description
Highly potent and selective 5-HT uptake inhibitor that binds with high affinity to the serotonin transporter (Ki = 0.05 nM). Ki values are 1.1, 350 and 1100 nM for inhibition of [3H]-5-HT, [3H]-l-NA and [3H]-DA uptake respectively. Displays minimal affinity for α1-, α2- or β-adrenoceptors, 5-HT2A, 5-HT1A, D2 or H1 receptors at concentrations below 1000 nM, however displays weak affinity for muscarinic ACh receptors (Ki = 42 nM). Antidepressant and anxiolytic in vivo.Specifications
>99% | |
Paroxetine maleate | |
445.44 | |
Paroxetine maleate |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title