Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patent Blue V calcium salt, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014008 Shop All MilliporeSigma Supelco Products
Click to view available options
:
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging

Bottomless Glass Bottle with Inserted Fused Cone

Quantity

25 mg

CAS

3536-49-0

Type

Analytical Standard

Linear Formula

C27H31N2O7S2 · 0.5 Ca

MDL Number

MFCD00038746

Synonym

Acid Blue 3 calcium salt; Acid Blue V calcium salt; Food Blue 5; Patent Blue V hemicalcium salt

Formula Weight

579.71

Percent Purity

≥97.0% (HPLC)

Grade

Analytical Standard

Physical Form

Neat

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.