Learn More
Pc2 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody
$154.46
Specifications
Antigen | Pc2 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2396
![]() |
Bethyl Laboratories, Inc
A302355AT |
20 μL |
Each of 1 for $154.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
CBX4 is a member of Drosophila Polycomb group gene family. The polycomb group (PcG) genes are essential for maintece of proper expression patterns of developmental master regulators; changes in expression pf PcG proteins have been associated with cancer. CBX4 is a part of the cellular memory system responsible for the inheritance of gene activity by progeny cells. It participates in maintaining the transcriptionally repressive state of genes. CBX4 is part of acomplex that acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. CBX4 is an E3 SUMO-protein ligase which facilitates SUMO1 conjugation by UBE2I.Specifications
Pc2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
E3 SUMO-protein ligase CBX4; Chromobox protein homolog 4; Polycomb 2 homolog; Pc2; hPc2 | |
Between 150 and 200 | |
Primary | |
4° C | |
CBX4 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O00257, O55187 | |
12418, 8535 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.