Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCI 34051, Tocris Bioscience™

Catalog No. p-4835954 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective HDAC8 inhibitor

Potent and selective histone deacetylase 8 (HDAC8) inhibitor (IC50 = 10 nM). Displays >200 fold selectivity over HDAC isoforms 1, 2, 3, 6 and 10. Induces apoptosis in cell lines derived from T cell lymphomas or leukemias.

Chemical Identifiers

CAS 950762-95-5
Molecular Formula C17H16N2O3
Molecular Weight (g/mol) 296.326
InChI Key AJRGHIGYPXNABY-UHFFFAOYSA-N
Synonym unii-pui52vxv61, n-hydroxy-1-4-methoxybenzyl-1h-indole-6-carboxamide, pui52vxv61, n-hydroxy-1-4-methoxyphenyl methyl indole-6-carboxamide, n-hydroxy-1-4-methoxyphenyl methyl-1h-indole-6-carboxamide, d0q5mc, 1h-indole-6-carboxamide, n-hydroxy-1-4-methoxyphenyl methyl, hdac 8 inhibitors cancer/inflammation, 1-4-methoxybenzyl-n-hydroxy-1h-indole-6-carboxamide, 1-4-methoxyphenyl methyl indole-6-carbohydroxamic acid
PubChem CID 24753719
IUPAC Name N-hydroxy-1-[(4-methoxyphenyl)methyl]indole-6-carboxamide
SMILES COC1=CC=C(C=C1)CN2C=CC3=C2C=C(C=C3)C(=O)NO

Specifications

Quantity 10 mg
Formula Weight 296.32
Chemical Name or Material PCI 34051
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.