Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD 0325901, Tocris Bioscience™

Catalog No. p-4817022 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent inhibitor of MEK1/2

PD 0325901 is a potent MEK1 and MEK2 inhibitor. PD 0325901 inhibits MEK activity in mouse colon 26 cells (IC50 = 0.33 nM). PD 0325901 inhibits the growth of melanoma cell lines in vitro and in vivo; induces G1-phase cell cycle arrest and apoptosis in a mouse xenograft model. Also inhibits production of proangiogenic cytokines such as VEGF. Enhances generation of induced pluripotent stem cells (iPSCs). Orally active.

For more information about how PD 0325901 may be used, see our protocols: Culturing Transcriptomically Distinct Pluripotent mESCs, Highly Efficient Generation of CiPSCs from MEFs, Accelerated Induction of Cortical Neurons from hiPSCs .

Chemical Identifiers

CAS 391210-10-9
Molecular Formula C16H14F3IN2O4
Molecular Weight (g/mol) 482.198
InChI Key SUDAHWBOROXANE-SECBINFHSA-N
Synonym r-n-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, r-n-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenylamino benzamide, unii-86k0j5ak6m, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-2r-2,3-dihydroxypropyl oxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-r-2,3-dihydroxy-propoxy-3,4-difluoro-2-2-fluoro-4-iodo-phenylamino-benzamide, n-r-2,3-dihydroxypropyl oxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodoanilino benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino-benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodo-anilino benzamide
PubChem CID 9826528
IUPAC Name N-[(2R)-2,3-dihydroxypropoxy]-3,4-difluoro-2-(2-fluoro-4-iodoanilino)benzamide
SMILES C1=CC(=C(C=C1I)F)NC2=C(C=CC(=C2F)F)C(=O)NOCC(CO)O

Specifications

Quantity 10 mg
Formula Weight 482.19
Percent Purity >99%
Chemical Name or Material PD 0325901
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.