Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-PD 128907 hydrochloride, Tocris Bioscience™

Catalog No. p-4783076 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity D3 agonist (D3 ≥ D2 > D4)

High affinity D3 dopamine receptor agonist (Ki = 2.3 nM). Displays 18 - 200-fold selectivity over other dopamine receptor subtypes.

Chemical Identifiers

CAS 300576-59-4
Molecular Formula C14H20ClNO3
Molecular Weight (g/mol) 285.77
MDL Number MFCD00210210,MFCD06858201
InChI Key DCFXOTRONMKUJB-UHFFFAOYNA-N
Synonym +/--pd 128,907 hydrochloride, s +-pd 128,907 hydrochloride, pd128907 hydrochloride, +-pd 128907 hydrochloride, +-pd 128,907 hydrochloride, rel-4ar,10br-4-propyl-2,3,4,4a,5,10b-hexahydrochromeno 4,3-b 1,4 oxazin-9-ol hydrochloride, +-4ar,10br-3,4,4a,10b-tetrahydro-4-propyl-2h,5h-1 benzopyrano 4,3-b-1,4-oxazin-9-ol hydrochloride, 4ar,10br-rel-4-propyl-2,3,4,4a,5,10b-hexahydrochromeno 4,3-b 1,4 oxazin-9-ol hydrochloride, 4ar,10br-4-propyl-3,4,4a,10b-tetrahydro-2h,5h-chromeno 4,3-b 1,4 oxazin-9-ol hydrochloride, y-pd 128,907 hydrochloride
PubChem CID 11957668
IUPAC Name 6-propyl-3,9-dioxa-6-azatricyclo[8.4.0.0²,⁷]tetradeca-1(10),11,13-trien-13-ol hydrochloride
SMILES Cl.CCCN1CCOC2C1COC1=C2C=C(O)C=C1

Specifications

Quantity 10 mg
Formula Weight 285.77
Percent Purity >99%
Chemical Name or Material (+)-PD 128907 hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.