Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD 146176, Tocris Bioscience™

Catalog No. p-4784246 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 15-lipoxygenase inhibitor

Specific, non-competitve 15-lipoxygenase (15-LOX) inhibitor (Ki = 197nM) that has no demonstrable effect on 5-LOX, 12-LOX, COX-1 or COX-2 (IC50 = 0.54 μM for 15-LOX in rabbit reticulocytes). Lacks non-specific antioxidant properties and prevents atherogenesis via regulation of monocyte-macrophage enrichment in vivo.

Chemical Identifiers

CAS 4079-26-9
Molecular Formula C15H11NS
Molecular Weight (g/mol) 237.32
InChI Key ZGOOPZVQMLHPFM-UHFFFAOYSA-N
Synonym 6,11-dihydrothiochromeno 4,3-b indole, 6,11-dihydro 1 benzothiopyrano 4,3-b indole, 6,3-b indole, d0j8rs, 6,11-dihydro-5-thia-11-aza-benzo a fluorene, 1 benzothiopyrano 4, 6,11-dihydro, 6h,11h-thiochromeno 4,3-b indole, pd hplc , solid, 1 benzothiopyrano 4,3-b indole, 6,11-dihydro, 8-thia-17-azatetracyclo 8.7.0.0 2 ,?.0 1 1 , 1 ? heptadeca-1 10 ,2 7 ,3,5,11,13,15-heptaene
PubChem CID 297589
IUPAC Name 6,11-dihydrothiochromeno[4,3-b]indole
SMILES C1C2=C(C3=CC=CC=C3S1)NC4=CC=CC=C24

Specifications

Quantity 10 mg
Formula Weight 237.32
Percent Purity >98%
Chemical Name or Material PD 146176
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.