Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD 168077 maleate, Tocris Bioscience™

Catalog No. p-4785323 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity, selective D4 agonist

A potent D4 dopamine receptor agonist (Ki = 8.7 nM) with > 400-fold selectivity over D2 and > 300-fold selectivity versus D3 subtypes respectively. Induces synaptic translocation of CaMK II to postsynaptic sites in cultured prefrontal cortical neurons. Centrally active in vivo.

Chemical Identifiers

CAS 630117-19-0
Molecular Formula C24H26N4O5
Molecular Weight (g/mol) 450.495
InChI Key NAEUGRPISCANHO-BTJKTKAUSA-N
Synonym pd 168077 maleate, pd 168,077 maleate, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide maleate, pd 168,077 maleate salt, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide; maleic acid, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methylbenzamide maleate salt, pd168077 maleate, pd 168,077 maleate salt, powder, n-methyl-4-2-cyanophenyl piperazinyl-3-methylbenzamide maleate, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methyl-benzamide maleate
PubChem CID 11957665
IUPAC Name (Z)-but-2-enedioic acid;N-[[4-(2-cyanophenyl)piperazin-1-yl]methyl]-3-methylbenzamide
SMILES CC1=CC=CC(=C1)C(=O)NCN2CCN(CC2)C3=CC=CC=C3C#N.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 450.49
Percent Purity 98%
Chemical Name or Material N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.