Learn More
PD-L1 (CD274) Mouse anti-Human, BV650, Clone: 29E.2A3, BD Horizon™
Mouse Monoclonal Antibody
$221.00 - $444.00
Specifications
Antigen | PD-L1 (CD274) |
---|---|
Clone | 29E.2A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 650 |
Description
PD-L1 (CD274) Monoclonal antibody specifically detects PD-L1 (CD274) in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
PD-L1 (CD274) | |
Flow Cytometry | |
Brilliant Violet 650 | |
Mouse | |
Human | |
B7H1; PD-L1; PDCD1L1; PDCD1LG1; PDL1; hPD-L1 | |
Human PD-L1 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
29E.2A3 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
29126 | |
IgG2b κ | |
Affinity Chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.