Learn More
ACROBiosystems ACROBiosystems Human PD-L1 / B7-H1 Protein, His Tag (HPLC verified)
Supplier:  ACROBiosystems PD1H52291MG
Encompass_Preferred
Recombinant Protein;1MG;Human PD-L1, His Tag (HPLC verified) (PD1-H5229) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1 ).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, His Tag (HPLC verified) (Cat. No. PD1-H5229) at 1 μg/mL (100 μL/well) can bind Anti-Human PD-L1 MAb (Human IgG1) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Human PD-L1, His Tag (HPLC verified) (Cat. No. PD1-H5229) at series of concentration on MonoRabᵀᴹ Anti-His Tag (C-term) Antibody precoated (0.1 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) at 5 μg/mL (100 μL/well) with a linear range of 10-78 ng/mL (Routinely tested).;Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) at 1 μg/mL (5 μL/well) can bind Human PD-L1, His Ta
Catalog No. 50-195-6669
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.