Learn More
Medchemexpress LLC HY-103318 2mg Medchemexpress, PD173212 CAS:217171-01-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033182MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-103318 2mg PD173212 CAS:217171-01-2 PD173212 is a selective N-type voltage sensitive calcium channel (VSCC) blocker, with an IC50 of 36 nM in IMR-32 assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8073
missing translation for 'QuantityLeft'