Learn More
Perkin Elmer US LLC Combustion Tube for CHN Analysis, Quartz
Supplier: Perkin Elmer US LLC N2411264
PerkinElmer combustion tubes are composed of pure fused quartz which is 99% Si02. Fused quartz has been chosen because of its very high purity and low thermal expansion rate. Because we care about your safety, all combustion tubes are pressure-tested to eliminate any tubes which might contain weak spots. In addition, stringent specifications are used to insure that the combustion tubes fit in an airtight connection when placed in the analyzer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.