Learn More
Restek™ PEEK Secure-Fit Fitting, Single Connector
Secure-Fit connectors ensure a consistent, leak-free seal and eliminate excess dead volume.
$69.74 - $69.76
Catalog Number | Mfr. No. | Diameter (English) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
06-712-784
![]() |
Restek™
25237 |
0.007 in. | 30 cm |
Each for $69.76
|
|
||||
06-712-783
![]() |
Restek™
25236 |
0.007 in. | 20 cm |
Each for $69.74
|
|
||||
Description
An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.- Material: PEEK™
Specifications
Connector | |
0.007 in. | |
PEEK |