Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ PEEK Tubing

Useful for when contact between the sample and metal components must be avoided in an HPLC system, such as when working with metal complexing agents or certain biochemical compounds

Supplier:  MilliporeSigma™ Supelco™ Z227293

 View more versions of this product

Catalog No. 11-101-1568


Description

Description

  • A popular replacement for stainless steel tubing in various places in the HPLC system
  • Useful when contact between the sample and metal components must be avoided, such as when working with metal complexing agents or certain biochemical compounds
  • Tubing is flexible, easy to cut, has excellent mechanical stability and chemical compatibility
  • Tetrahydrofuran (THF), dimethyl sulfoxide (DMSO), methylene chloride, and concentrated nitric and sulfuric acids should not be used with PEEK tubing
  • 1/16 in. tubing is color coded for easy identification of the internal diameter
  • Maximum Temperature: 100°C
Specifications

Specifications

Tubing
HPLC Systems
1/16 in.
3 m
6000 psi
Orange
0.020 in.
10 ft.
PEEK
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.