Learn More
MilliporeSigma™ Supelco™ PEEKsil™ Tubing
Fused silica tubing sheathed in PEEK polymer, with an effective outer diameter
Supplier:  MilliporeSigma™ Supelco™ 51319U
 View more versions of this product
Catalog No. 11-101-1744
Explore all special offers
Add to Cart
Description
Description
  • PEEK sheating is mechanically strong and has ideal characteristics for sealing with metal or polymer fittings
  • Fused silica core provides a consistent and rigid fluid pathway with very tight tolerances and industry-accepted chemical properties
  • Extremely low absorption characteristics, especially when compared to stainless steel
  • Excellent chemical compatibility
  • Note: Do not cut this tubing - it should be used as its precut lengths because of permanent damage caused by conventional cutters
Specifications
Specifications
Tubing
HPLC
1/32 in.
0.8 mm
Polymer-Sheathed Fused Silica
Natural
0.002 in.
50 µm
10 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
PEEKSil is a trademark of SGE Analytical Science Pty Ltd.